Order Entry
Poland
ContactUsLinkComponent
Recombinat Protein,Human TrkB NTRK2 Protein, His Tag,Human TrkB, His Tag (NT2-H5228) is expressed from human 293 cells (HEK293). It contains AA Cys 32 - His 430 (Accession AAH31835).This protein carries a polyhistidine tag at the C-terminus. The prot 1 * 1 mg
Catalog # ACRBNT2-H5228-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Human TrkB NTRK2 Protein, His Tag,Human TrkB, His Tag (NT2-H5228) is expressed from human 293 cells (HEK293). It contains AA Cys 32 - His 430 (Accession AAH31835).This protein carries a polyhistidine tag at the C-terminus. The prot 1 * 1 mg
Catalog # ACRBNT2-H5228-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together