Order Entry
Poland
ContactUsLinkComponent
Mouse CD40, His Tag (TN5-M52H8) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Arg 193 (Accession P27512-1). 1 * 100 µG
Catalog # ACRBTN5-M52H8100UG
Supplier:  ACRO Biosystems
Mouse CD40, His Tag (TN5-M52H8) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Arg 193 (Accession P27512-1). 1 * 100 µG
Catalog # ACRBTN5-M52H8100UG
Supplier:  ACRO Biosystems
Frequently Bought Together