Order Entry
Poland
ContactUsLinkComponent
Biotinylated Human TACI, Fc,Avitag (TAI-H82F6) is expressed from human 293 cells (HEK293). It contains AA Ser 2 - Thr 166 (Accession O14836-1). 1 * 200 µG
  ACRBTAI-H82F6200UG
Biotinylated Human TACI, Fc,Avitag (TAI-H82F6) is expressed from human 293 cells (HEK293). It contains AA Ser 2 - Thr 166 (Accession O14836-1). 1 * 200 µG
  ACRBTAI-H82F6200UG
Frequently Bought Together