Order Entry
Poland
ContactUsLinkComponent
Recombinat Protein,Human Serpin E2 PN1 Protein, His Tag,Human Serpin E2, His Tag (SE2-H5225) is expressed from human 293 cells (HEK2 93). It contains AA Ser 20 - Pro 397 (Accession P07093-2).This protein carries a polyhistidine tag at the C-terminus. 1 * 25 µG
  ACRBSE2-H5225-25UG
Recombinat Protein,Human Serpin E2 PN1 Protein, His Tag,Human Serpin E2, His Tag (SE2-H5225) is expressed from human 293 cells (HEK2 93). It contains AA Ser 20 - Pro 397 (Accession P07093-2).This protein carries a polyhistidine tag at the C-terminus. 1 * 25 µG
  ACRBSE2-H5225-25UG
Frequently Bought Together