Order Entry
Poland
ContactUsLinkComponent
Recombinat Protein,Human Siglec-8 Protein, His Tag,Human Siglec-8, His Tag (SI8-H52H8) is expressed from human 293 cells (HEK293). I t contains AA Met 17 - Ala 363 (Accession Q9NYZ4-1).This protein carries a polyhistidine tag at the C-terminus. The p 1 * 100 µG
  ACRBSI8-H52H8100UG
Recombinat Protein,Human Siglec-8 Protein, His Tag,Human Siglec-8, His Tag (SI8-H52H8) is expressed from human 293 cells (HEK293). I t contains AA Met 17 - Ala 363 (Accession Q9NYZ4-1).This protein carries a polyhistidine tag at the C-terminus. The p 1 * 100 µG
  ACRBSI8-H52H8100UG
Frequently Bought Together