Order Entry
Poland
ContactUsLinkComponent
Recombinat Protein,Human PH20 SPAM1 Protein, His Tag,Human PH20, His Tag (PH0-H5225) is expressed from human 293 cells (HEK293). It contains AA Leu 36 - Tyr 482 (Accession AAH26163).This protein carries a polyhistidine tag at the C-terminus. The prot 1 * 1 mg
Catalog # ACRBPH0-H5225-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Human PH20 SPAM1 Protein, His Tag,Human PH20, His Tag (PH0-H5225) is expressed from human 293 cells (HEK293). It contains AA Leu 36 - Tyr 482 (Accession AAH26163).This protein carries a polyhistidine tag at the C-terminus. The prot 1 * 1 mg
Catalog # ACRBPH0-H5225-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together