Order Entry
Poland
ContactUsLinkComponent
Recombinat Protein,Human MERTK Mer Protein, His Tag,Human MERTK, His Tag (MEK-H52H6) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Ile 505 (Accession Q12866-1).This protein carries a polyhistidine tag at the C-terminus. The prot 1 * 100 µG
Catalog # ACRBMEK-H52H6100UG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human MERTK Mer Protein, His Tag,Human MERTK, His Tag (MEK-H52H6) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Ile 505 (Accession Q12866-1).This protein carries a polyhistidine tag at the C-terminus. The prot 1 * 100 µG
Catalog # ACRBMEK-H52H6100UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together