Order Entry
Poland
ContactUsLinkComponent
Human GM-CSF R alpha, Fc Tag (GRA-H5255) is expressed from human 293 cells (HEK293). It contains AA Glu 23 - Gly 320 (Accession P155 09-1). 1 * 100 µG
Catalog # ACRBGRA-H5255100UG
Supplier:  ACRO Biosystems
Human GM-CSF R alpha, Fc Tag (GRA-H5255) is expressed from human 293 cells (HEK293). It contains AA Glu 23 - Gly 320 (Accession P155 09-1). 1 * 100 µG
Catalog # ACRBGRA-H5255100UG
Supplier:  ACRO Biosystems
Frequently Bought Together