Order Entry
Poland
ContactUsLinkComponent
Recombinat Protein,Human NME1 NDKA NM23-H1 Protein, His Tag,Human NME1, His Tag (NM1-H5147) is expressed from E.coli cells. It conta ins AA Ala 2 - Glu 152 (Accession AAH00293).This protein carries a polyhistidine tag at the N-terminus. The protein h 1 * 1 mg
Catalog # ACRBNM1-H5147-1MG
Dostawca:  ACRO Biosystems
Recombinat Protein,Human NME1 NDKA NM23-H1 Protein, His Tag,Human NME1, His Tag (NM1-H5147) is expressed from E.coli cells. It conta ins AA Ala 2 - Glu 152 (Accession AAH00293).This protein carries a polyhistidine tag at the N-terminus. The protein h 1 * 1 mg
Catalog # ACRBNM1-H5147-1MG
Dostawca:  ACRO Biosystems
Frequently Bought Together