Order Entry
Poland
ContactUsLinkComponent
Recombinat Protein,Human ALCAM Protein, His Tag,Human ALCAM, His Tag (CD6-H5228) is expressed from human 293 cells (HEK293). It cont ains AA Trp 28 - Ala 526 (Accession Q13740-1).This protein carries a polyhistidine tag at the C-terminus. The protein 1 * 1 mg
Catalog # ACRBCD6-H5228-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Human ALCAM Protein, His Tag,Human ALCAM, His Tag (CD6-H5228) is expressed from human 293 cells (HEK293). It cont ains AA Trp 28 - Ala 526 (Accession Q13740-1).This protein carries a polyhistidine tag at the C-terminus. The protein 1 * 1 mg
Catalog # ACRBCD6-H5228-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together