Order Entry
Poland
ContactUsLinkComponent
Human TRAIL R2, His Tag (TR2-H5229) is expressed from human 293 cells (HEK293). It contains AA Ile 56 - Glu 182 (Accession NP003833) . 1 * 100 µG
Catalog # ACRBTR2-H5229100UG
Supplier:  ACRO Biosystems
Human TRAIL R2, His Tag (TR2-H5229) is expressed from human 293 cells (HEK293). It contains AA Ile 56 - Glu 182 (Accession NP003833) . 1 * 100 µG
Catalog # ACRBTR2-H5229100UG
Supplier:  ACRO Biosystems
Frequently Bought Together