Order Entry
Poland
ContactUsLinkComponent
Biotinylated Human CD47, Fc (L234A, L235A, P329G),Avitag (CD7-H82F8) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Pro 139 (Accession Q08722-3). 1 * 200 µG
Catalog # ACRBCD7-H82F8200UG
Supplier:  ACRO Biosystems
Biotinylated Human CD47, Fc (L234A, L235A, P329G),Avitag (CD7-H82F8) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Pro 139 (Accession Q08722-3). 1 * 200 µG
Catalog # ACRBCD7-H82F8200UG
Supplier:  ACRO Biosystems
Frequently Bought Together