Order Entry
Poland
ContactUsLinkComponent
Recombinat Protein,Human SMAC Diablo Protein, His Tag,Human SMAC, His Tag (DIO-H5129) is expressed from E.coli cells. It contains AA Ala 56 - Asp 239 (Accession NP063940).This protein carries a polyhistidine tag at the C-terminus. The protein has a f 1 * 200 µG
Catalog # ACRBDIO-H5129200UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human SMAC Diablo Protein, His Tag,Human SMAC, His Tag (DIO-H5129) is expressed from E.coli cells. It contains AA Ala 56 - Asp 239 (Accession NP063940).This protein carries a polyhistidine tag at the C-terminus. The protein has a f 1 * 200 µG
Catalog # ACRBDIO-H5129200UG
Supplier:  ACRO Biosystems
Frequently Bought Together