Order Entry
Poland
ContactUsLinkComponent
Cynomolgus Transferrin R, His Tag (TFR-C524a) is expressed from human 293 cells (HEK293). It contains AA Cys 89 - Phe 760 (Accession XP005545315.1). 1 * 1 mg
Catalog # ACRBTFR-C524A-1MG
Supplier:  ACRO Biosystems
Cynomolgus Transferrin R, His Tag (TFR-C524a) is expressed from human 293 cells (HEK293). It contains AA Cys 89 - Phe 760 (Accession XP005545315.1). 1 * 1 mg
Catalog # ACRBTFR-C524A-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together