Order Entry
Poland
ContactUsLinkComponent
Recombinat Protein,Human HSP90AA1 HSP86 Protein, GST Tag,Human HSP90AA1, GST Tag (HS1-H513x) is expressed from E.coli cells. It cont ains AA Glu 535 - Asp 732 (Accession NP005339).This protein carries a GST tag at the N-terminus. The protein has a ca 1 * 1 mg
Catalog # ACRBHS1-H513X-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Human HSP90AA1 HSP86 Protein, GST Tag,Human HSP90AA1, GST Tag (HS1-H513x) is expressed from E.coli cells. It cont ains AA Glu 535 - Asp 732 (Accession NP005339).This protein carries a GST tag at the N-terminus. The protein has a ca 1 * 1 mg
Catalog # ACRBHS1-H513X-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together