Order Entry
Poland
ContactUsLinkComponent
Biotinylated Human CD155, Fc,Avitag (CD5-H82F6) is expressed from human 293 cells (HEK293). It contains AA Trp 21 - Asn 343 (Accessi on NP006496.2). 1 * 25 µG
  ACRBCD5-H82F6-25UG
Biotinylated Human CD155, Fc,Avitag (CD5-H82F6) is expressed from human 293 cells (HEK293). It contains AA Trp 21 - Asn 343 (Accessi on NP006496.2). 1 * 25 µG
  ACRBCD5-H82F6-25UG
Frequently Bought Together