Order Entry
Poland
ContactUsLinkComponent
Recombinat Protein,Human LBP Lipopolysaccharide Protein, His Tag,Human Lipopolysaccharide, His Tag (LBP-H52H5) is expressed from hum an 293 cells (HEK293). It contains AA Ala 26 - Val 481 (Accession P18428).This protein carries a polyhistidine tag at 1 * 100 µG
Catalog # ACRBLBP-H52H5100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human LBP Lipopolysaccharide Protein, His Tag,Human Lipopolysaccharide, His Tag (LBP-H52H5) is expressed from hum an 293 cells (HEK293). It contains AA Ala 26 - Val 481 (Accession P18428).This protein carries a polyhistidine tag at 1 * 100 µG
Catalog # ACRBLBP-H52H5100UG
Supplier:  ACRO Biosystems
Frequently Bought Together