Order Entry
Poland
ContactUsLinkComponent
Recombinat Protein,Human Stathmin 1 STMN1 Protein, His Tag,Human Stathmin 1, His Tag (ST1-H5149) is expressed from E.coli cells. It contains AA Ala 2 - Asp 149 (Accession NP981946).This protein carries a polyhistidine tag at the N-terminus. The prote 1 * 1 mg
Catalog # ACRBST1-H5149-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Human Stathmin 1 STMN1 Protein, His Tag,Human Stathmin 1, His Tag (ST1-H5149) is expressed from E.coli cells. It contains AA Ala 2 - Asp 149 (Accession NP981946).This protein carries a polyhistidine tag at the N-terminus. The prote 1 * 1 mg
Catalog # ACRBST1-H5149-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together