Order Entry
Poland
ContactUsLinkComponent
Recombinat Protein,Human LYPD3 C4.4A Protein, His Tag,Human LYPD3, His Tag (LY3-H52H5) is expressed from human 293 cells (HEK293). I t contains AA Leu 31 - His 286 (Accession AAH39167).This protein carries a polyhistidine tag at the C-terminus. The p 1 * 100 µG
Catalog # ACRBLY3-H52H5100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human LYPD3 C4.4A Protein, His Tag,Human LYPD3, His Tag (LY3-H52H5) is expressed from human 293 cells (HEK293). I t contains AA Leu 31 - His 286 (Accession AAH39167).This protein carries a polyhistidine tag at the C-terminus. The p 1 * 100 µG
Catalog # ACRBLY3-H52H5100UG
Supplier:  ACRO Biosystems
Frequently Bought Together