Order Entry
Poland
ContactUsLinkComponent
FITC-Labeled Human Glypican 3, His Tag (Cat. No. GP3-HF2H1) is expressed from human HEK293 cells. It contains AA Gln 25 - His 559 (A ccession P51654-1). It is the FITC labeled form of Human Glypican 3, His Tag (Cat. No. GP3-H52H4). 1 * 200 µG
Catalog # ACRBGP3-HF2H1200UG
Supplier:  ACRO Biosystems
FITC-Labeled Human Glypican 3, His Tag (Cat. No. GP3-HF2H1) is expressed from human HEK293 cells. It contains AA Gln 25 - His 559 (A ccession P51654-1). It is the FITC labeled form of Human Glypican 3, His Tag (Cat. No. GP3-H52H4). 1 * 200 µG
Catalog # ACRBGP3-HF2H1200UG
Supplier:  ACRO Biosystems
Frequently Bought Together