Order Entry
Poland
ContactUsLinkComponent
Recombinat Protein,Biotinylated Human MIF Protein, His,Avitag™,Biotinylated Human MIF, His,Avitag (MIF-H82E7) is expressed from huma n 293 cells (HEK293). It contains AA Pro 2 - Ala 115 (Accession P14174-1).This protein carries a polyhistidine tag at 1 * 25 µG
  ACRBMIF-H82E7-25UG
Recombinat Protein,Biotinylated Human MIF Protein, His,Avitag™,Biotinylated Human MIF, His,Avitag (MIF-H82E7) is expressed from huma n 293 cells (HEK293). It contains AA Pro 2 - Ala 115 (Accession P14174-1).This protein carries a polyhistidine tag at 1 * 25 µG
  ACRBMIF-H82E7-25UG
Frequently Bought Together