Order Entry
Poland
ContactUsLinkComponent
Recombinat Protein,Human Jagged 1 JAG1 Protein, His Tag,Human Jagged 1, His Tag (JA1-H52H9) is expressed from human 293 cells (HEK29 3). It contains AA Gln 34 - Ser 1046 (Accession P78504-1).This protein carries a polyhistidine tag at the C-terminus. 1 * 1 mg
  ACRBJA1-H52H9-1MG
Recombinat Protein,Human Jagged 1 JAG1 Protein, His Tag,Human Jagged 1, His Tag (JA1-H52H9) is expressed from human 293 cells (HEK29 3). It contains AA Gln 34 - Ser 1046 (Accession P78504-1).This protein carries a polyhistidine tag at the C-terminus. 1 * 1 mg
  ACRBJA1-H52H9-1MG
Frequently Bought Together