Order Entry
Poland
ContactUsLinkComponent
Recombinat Protein,Cynomolgus CD38 Protein, His Tag,Cynomolgus CD38, His Tag (CD8-C5223) is expressed from human 293 cells (HEK293). It contains AA Leu 44 - Ile 301 (Accession Q5VAN0-1).This protein carries a polyhistidine tag at the C-terminus. The 1 * 100 µG
  ACRBCD8-C5223100UG
Recombinat Protein,Cynomolgus CD38 Protein, His Tag,Cynomolgus CD38, His Tag (CD8-C5223) is expressed from human 293 cells (HEK293). It contains AA Leu 44 - Ile 301 (Accession Q5VAN0-1).This protein carries a polyhistidine tag at the C-terminus. The 1 * 100 µG
  ACRBCD8-C5223100UG
Frequently Bought Together