Order Entry
Poland
ContactUsLinkComponent
Recombinat Protein,Human Galectin-3 Protein, His Tag,Human Galectin-3, His Tag (GA3-H5129) is expressed from E.coli cells. It contai ns AA Met 1 - Ile 250 (Accession NP002297).This protein carries a polyhistidine tag at the C-terminus. The protein ha 1 * 200 µG
Catalog # ACRBGA3-H5129200UG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human Galectin-3 Protein, His Tag,Human Galectin-3, His Tag (GA3-H5129) is expressed from E.coli cells. It contai ns AA Met 1 - Ile 250 (Accession NP002297).This protein carries a polyhistidine tag at the C-terminus. The protein ha 1 * 200 µG
Catalog # ACRBGA3-H5129200UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together