Order Entry
Poland
ContactUsLinkComponent
FITC-Labeled Human Glypican 3, Fc Tag (Cat. No. GP3-HF258) is expressed from human HEK293 cells. It contains AA Gln 25 - His 559 (Ac cession P51654-1). It is the FITC labeled form of Human Glypican 3, Fc Tag (Cat. No. GP3-H5258). 1 * 25 µG
Catalog # ACRBGP3-HF258-25UG
Supplier:  ACRO Biosystems
FITC-Labeled Human Glypican 3, Fc Tag (Cat. No. GP3-HF258) is expressed from human HEK293 cells. It contains AA Gln 25 - His 559 (Ac cession P51654-1). It is the FITC labeled form of Human Glypican 3, Fc Tag (Cat. No. GP3-H5258). 1 * 25 µG
Catalog # ACRBGP3-HF258-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together