Order Entry
Poland
ContactUsLinkComponent
FITC-Labeled Human Siglec-2, His Tag (Cat. No. SI2-HF2H6) is expressed from human HEK293 cells. It contains AA Asp 20 - Arg 687 (Acc ession P20273-1). It is the FITC labeled form of Human Siglec-2, His Tag (Cat. No. CD2-H52H8). 1 * 200 µG
Catalog # ACRBSI2-HF2H6200UG
Supplier:  ACRO Biosystems
FITC-Labeled Human Siglec-2, His Tag (Cat. No. SI2-HF2H6) is expressed from human HEK293 cells. It contains AA Asp 20 - Arg 687 (Acc ession P20273-1). It is the FITC labeled form of Human Siglec-2, His Tag (Cat. No. CD2-H52H8). 1 * 200 µG
Catalog # ACRBSI2-HF2H6200UG
Supplier:  ACRO Biosystems
Frequently Bought Together