Order Entry
Poland
ContactUsLinkComponent
Biotinylated Human IL-17AIL-17F, Avitag,His Tag (ILF-H82W1) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Ala 155 (IL-17A) Arg 31 - Gln 163 (IL-17F) (Accession Q16552-1 (IL-17A) Q96PD4-1 (IL-17F)). 1 * 25 µG
Catalog # ACRBILF-H82W1-25UG
Supplier:  ACROBIOSYSTEMS
Biotinylated Human IL-17AIL-17F, Avitag,His Tag (ILF-H82W1) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Ala 155 (IL-17A) Arg 31 - Gln 163 (IL-17F) (Accession Q16552-1 (IL-17A) Q96PD4-1 (IL-17F)). 1 * 25 µG
Catalog # ACRBILF-H82W1-25UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together