Order Entry
Poland
ContactUsLinkComponent
Human IL-33, His Tag (IL3-H52H7) is expressed from human 293 cells (HEK293). It contains AA His 109 - Thr 270 (Accession O95760-1). 1 * 50 µG
Catalog # ACRBIL3-H52H7-50UG
Supplier:  ACRO Biosystems
Human IL-33, His Tag (IL3-H52H7) is expressed from human 293 cells (HEK293). It contains AA His 109 - Thr 270 (Accession O95760-1). 1 * 50 µG
Catalog # ACRBIL3-H52H7-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together