Order Entry
Poland
ContactUsLinkComponent
Recombinat Protein,Human MBL Protein, His Tag,Human MBL, His Tag (MBL-H5220) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Ile 248 (Accession AAH96179).This protein carries a polyhistidine tag at the C-terminus. The protein has 1 * 1 mg
Catalog # ACRBMBL-H5220-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Human MBL Protein, His Tag,Human MBL, His Tag (MBL-H5220) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Ile 248 (Accession AAH96179).This protein carries a polyhistidine tag at the C-terminus. The protein has 1 * 1 mg
Catalog # ACRBMBL-H5220-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together