Recombinat Protein,Cynomolgus TSLP (R127A, R130S) Protein, His Tag,Cynomolgus TSLP (R127A, R130S), His Tag (TSP-C52H4) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Accession A0A2K5TXV0-1 (R127A, R130S)).This protein ca 1 * 100 µG
Catalog # ACRBTSP-C52H4100UG
Supplier: ACROBIOSYSTEMS
Frequently Bought Together