Order Entry
Poland
ContactUsLinkComponent
Recombinat Protein,Cynomolgus IL-3 R alpha CD123 Protein, His Tag,Cynomolgus IL-3 R alpha, His Tag (ILA-C52H6) is expressed from hum an 293 cells (HEK293). It contains AA Arg 18 - Arg 302 (Accession G8F3K3-1).This protein carries a polyhistidine tag 1 * 100 µG
Catalog # ACRBILA-C52H6100UG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Cynomolgus IL-3 R alpha CD123 Protein, His Tag,Cynomolgus IL-3 R alpha, His Tag (ILA-C52H6) is expressed from hum an 293 cells (HEK293). It contains AA Arg 18 - Arg 302 (Accession G8F3K3-1).This protein carries a polyhistidine tag 1 * 100 µG
Catalog # ACRBILA-C52H6100UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together