Order Entry
Poland
ContactUsLinkComponent
Human FGF acidic, Tag Free (AFF-H4116) is expressed from E.coli cells. It contains AA Phe 16 - Asp 155 (Accession P05230-1). 1 * 200 µG
Catalog # ACRBAFF-H4116200UG
Supplier:  ACROBIOSYSTEMS
Human FGF acidic, Tag Free (AFF-H4116) is expressed from E.coli cells. It contains AA Phe 16 - Asp 155 (Accession P05230-1). 1 * 200 µG
Catalog # ACRBAFF-H4116200UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together