Order Entry
Poland
ContactUsLinkComponent
Recombinat Protein,Human PSGL-1 CD162 Protein, His Tag,Human PSGL-1, His Tag (PS1-H52H9) is expressed from human 293 cells (HEK293). It contains AA Gln 42 - Cys 320 (Accession Q14242-1).This protein carries a polyhistidine tag at the C-terminus. The 1 * 1 mg
Catalog # ACRBPS1-H52H9-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Human PSGL-1 CD162 Protein, His Tag,Human PSGL-1, His Tag (PS1-H52H9) is expressed from human 293 cells (HEK293). It contains AA Gln 42 - Cys 320 (Accession Q14242-1).This protein carries a polyhistidine tag at the C-terminus. The 1 * 1 mg
Catalog # ACRBPS1-H52H9-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together