Order Entry
Poland
ContactUsLinkComponent
Human FGL1, His Tag (FG1-H52Hy) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Ile 312 (Accession Q08830-1). 1 * 100 µG
Catalog # ACRBFG1-H52HY100UG
Supplier:  ACROBIOSYSTEMS
Human FGL1, His Tag (FG1-H52Hy) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Ile 312 (Accession Q08830-1). 1 * 100 µG
Catalog # ACRBFG1-H52HY100UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together