Order Entry
Poland
ContactUsLinkComponent
Recombinat Protein,Mouse RENIN Protein, His Tag,Mouse RENIN, His Tag (REN-M5222) is expressed from human 293 cells (HEK293). It cont ains AA Leu 22 - Arg 402 (Accession NP112469.1).This protein carries a polyhistidine tag at the C-terminus. The prote 1 * 1 mg
Catalog # ACRBREN-M5222-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Mouse RENIN Protein, His Tag,Mouse RENIN, His Tag (REN-M5222) is expressed from human 293 cells (HEK293). It cont ains AA Leu 22 - Arg 402 (Accession NP112469.1).This protein carries a polyhistidine tag at the C-terminus. The prote 1 * 1 mg
Catalog # ACRBREN-M5222-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together