Order Entry
Poland
ContactUsLinkComponent
Recombinat Protein,Human RENIN Protein, His Tag,Human RENIN, His Tag (REN-H5221) is expressed from human 293 cells (HEK293). It cont ains AA Leu 24 - Arg 406 (Accession AAH33474).This protein carries a polyhistidine tag at the C-terminus. The protein 1 * 1 mg
Catalog # ACRBREN-H5221-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Human RENIN Protein, His Tag,Human RENIN, His Tag (REN-H5221) is expressed from human 293 cells (HEK293). It cont ains AA Leu 24 - Arg 406 (Accession AAH33474).This protein carries a polyhistidine tag at the C-terminus. The protein 1 * 1 mg
Catalog # ACRBREN-H5221-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together