MABSol® Biotinylated Mouse TNF-alpha, His,Avitag (active trimer) (MALS verified) (TNA-M82E9) is expressed from human HEK293 cells. I t contains AA Leu 80 - Leu 235 (Accession P06804). 1 * 25 µG
Catalog # ACRBTNA-M82E9-25UG
Supplier: ACRO Biosystems
Frequently Bought Together