UTILITY FLOOR TOOL, Ø1.5 inch(Ø38mm) x 12 inch(30cm), STATIC CONDUCTIVE PLASTIC w/CONDUCTIVE NYLON BRISTLES 1 * 1 items
Catalog # TIGV381448A
Supplier: TIGER-VAC
UTILITY FLOOR TOOL, Ø1.5 inch(Ø38mm) x 12 inch(30cm), STATIC CONDUCTIVE PLASTIC w/CONDUCTIVE NYLON BRISTLES 1 * 1 items
Catalog # TIGV381448A
Supplier: TIGER-VAC
Frequently Bought Together
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Facial Tissues
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection