ETER NAFTOWY T.W. 40-60°C CZDA-BASIC OP. 1 * 1 L
Catalog # POCHBA4690115-1L
Supplier: POCH AVANTOR BRAND
ETER NAFTOWY T.W. 40-60°C CZDA-BASIC OP. 1 * 1 L
Catalog # POCHBA4690115-1L
Supplier: POCH AVANTOR BRAND
Frequently Bought Together
Aceton ≥99%, TECHNICAL
Supplier: VWR Chemicals
Aceton ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetonitryl ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Acetonitryl, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Woda, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Metanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Woda, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Woda, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetonitryl ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Aceton ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Eter naftowy, 40…60 °C, czysty reagent analityczny
Supplier: POCH AVANTOR BRAND
Etanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals