ACETON CZDA-BASIC 99,5% OP. 10 DM3 1 * 10 L
Catalog # POCHBA2480111-10L
Supplier: POCH AVANTOR BRAND
ACETON CZDA-BASIC 99,5% OP. 10 DM3 1 * 10 L
Catalog # POCHBA2480111-10L
Supplier: POCH AVANTOR BRAND
Frequently Bought Together
Woda, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Aceton ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Aceton ≥99%, TECHNICAL
Supplier: VWR Chemicals
Aceton ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Smoczki do pipet
Supplier: DEUTSCH NEUMANN
Acetonitryl ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Metanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetonitryl, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Woda, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Woda, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Strzykawki jednorazowego użytku, 3-częściowe, HENKE-JECT®, Henke Sass Wolf
Supplier: HENKE SASS WOLF
Acetonitryl ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Etanol, GPR RECTAPUR®
Supplier: VWR Chemicals
Etanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals