DI-SODU WERSENIAN 2 . HYDRAT (DISODU EDE 1 * 100 g
Catalog # POCH879810112-100G
Supplier: POCH AVANTOR BRAND
DI-SODU WERSENIAN 2 . HYDRAT (DISODU EDE 1 * 100 g
Catalog # POCH879810112-100G
Supplier: POCH AVANTOR BRAND
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
n-Hexane, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Toluene, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethyl acetate ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Methanol, EMPLURA®, Supelco®
Supplier: Merck
Toluene, EMPLURA®, extra pure, Supelco®
Supplier: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Water for TOC, Supelco®
Supplier: Merck
Potassium hydroxide 0.1 mol/l (0.1 N) in ethanol
Supplier: CHEMPUR
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Pyridine, extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific