SODU WODOROTLENEK MIKROGRANULKI CZDA OP. 1 * 500 g
Catalog # POCH810981118-500G
Supplier: POCH AVANTOR BRAND
SODU WODOROTLENEK MIKROGRANULKI CZDA OP. 1 * 500 g
Catalog # POCH810981118-500G
Supplier: POCH AVANTOR BRAND
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
ETYLOWY ALKOHOL 96% CZDA 1 * 500 mL
Supplier: POCH AVANTOR BRAND
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Hydrochloric acid 0.5 mol/l (0.5 N), AVS TITRINORM Reag. Ph. Eur. volumetric solution
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
TOLUEN CZDA, ODCZ. FP OP. 2,5 DM3 1 * 2,5 L
Supplier: POCH AVANTOR BRAND
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals