POTASU NADSIARCZAN CZDA OP. 100 G 1 * 100 g
Catalog # POCH743970111-100G
Supplier: POCH AVANTOR BRAND
POTASU NADSIARCZAN CZDA OP. 100 G 1 * 100 g
Catalog # POCH743970111-100G
Supplier: POCH AVANTOR BRAND
Frequently Bought Together
Potassium nitrate, Suprapur® for trace analysis, Supelco®
Supplier: Merck
Potassium peroxodisulphate ≥98.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Ammonium iron(II) sulphate hexahydrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Magnesium sulphate heptahydrate ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium chloride ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Sodium cyanide, Supelco®
Supplier: Merck
Sodium carbonate ≥97%, TECHNICAL
Supplier: VWR Chemicals
Disodium oxalate ≥99%
Supplier: Thermo Fisher Scientific
Tin(II) chloride dihydrate 98.0-103.0% ACS
Supplier: Thermo Fisher Scientific
Lithium chloride ≥98%, purified
Supplier: VWR Chemicals
Sodium chloride ≥99.5%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
Supplier: Honeywell Chemicals
Aluminium sulphate hydrate 98.0-102% ACS
Supplier: Thermo Fisher Scientific