KWAS SOLNY 1 MOL/L R-R (1 N) R-R MIANOWA 1 * 1 L
Catalog # POCH575320161-1L
Supplier: POCH AVANTOR BRAND
KWAS SOLNY 1 MOL/L R-R (1 N) R-R MIANOWA 1 * 1 L
Catalog # POCH575320161-1L
Supplier: POCH AVANTOR BRAND
Frequently Bought Together
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Titrants and reagents for volumetric Karl Fischer titrations, HYDRANAL™, Fluka™
Supplier: Honeywell Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Potassium dihydrogen phosphate ≥99.5%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Spoons with spatula end
Supplier: RSG
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Hydrochloric acid 34%, NORMATOM® for trace metal analysis
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
DI-SODU WERSENIAN 0,1 MOL/L R-R MIANOWAN 1 * 5 L
Supplier: POCH AVANTOR BRAND
Dialysis tubing, trial kits
Supplier: Spectrum Laboratories
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals