KWAS SIARKOWY (VI) 0,5 MOL/L (1 N) R-R M 1 * 1 L
Catalog # POCH575065166-1L
Supplier: POCH AVANTOR BRAND
KWAS SIARKOWY (VI) 0,5 MOL/L (1 N) R-R M 1 * 1 L
Catalog # POCH575065166-1L
Supplier: POCH AVANTOR BRAND
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Test kits, photometric, Chloride, LCK cuvette tests
Supplier: Hach
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Hydrochloric acid 0.5 mol/l (0.5 N), AVS TITRINORM Reag. Ph. Eur. volumetric solution
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Methanol, pure analytical reagent
Supplier: CHEMPUR
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection
Potassium fluoride, pure
Supplier: CHEMPUR