ANTYMONYLU POTASU WINIAN 3 . HYDRAT CZDA 1 * 100 g
Catalog # POCH148560110-100G
Supplier: POCH AVANTOR BRAND
ANTYMONYLU POTASU WINIAN 3 . HYDRAT CZDA 1 * 100 g
Catalog # POCH148560110-100G
Supplier: POCH AVANTOR BRAND
Frequently Bought Together
Methanol, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Water, TECHNICAL, demineralised
Supplier: VWR Chemicals
Benzyl alcohol, EMSURE® for analysis, Supelco®
Supplier: Merck
4-Aminobenzoic acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Acetic anhydride for synthesis, Sigma-Aldrich®
Supplier: Merck
Methanol, Prepsolv® for preparative chromatography, Supelco®
Supplier: Merck
Water, GPR RECTAPUR®
Supplier: VWR Chemicals
1-Propanol 99+%, extra pure
Supplier: Thermo Fisher Scientific
Ethanol absolute ≥99.9% (by GC), SPECTRONORM® for spectroscopy
Supplier: VWR Chemicals
n-Hexane ≥97%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Formaldehyde solution 35% (w/w) stabilised, meets analytical specification of BP, Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol, pure
Supplier: CHEMPUR
1-Pentanol 99+% ACS
Supplier: Thermo Fisher Scientific
Dichloromethane ≥99.8% stabilised, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
N,N-Dimethylformamide ≥99.8% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Dibutylamine for HPLC
Supplier: Thermo Fisher Scientific