ANTYMONYLU POTASU WINIAN 0.5 HYDRAT CZDA 1 * 10 g
CHMP111485604.0010
: CHEMPUR
Frequently Bought Together
Xylene - Mixture of isomers technical grade 1 * 5 L
: PanReac AppliChem
Acetonitrile ≥99.5% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
Ethyl acetate ≥99%, GPR RECTAPUR®
: VWR Chemicals
TERPENTYNA TECHN OP. 1 DM3 1 * 1 L
: POCH AVANTOR BRAND
Cyclohexanone ≥99.0% analytical reagent
: VWR Chemicals
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
: MACRON AVANTOR BRAND
N,N-Dimethylformamide, extra dry over Molecular Sieve 99.8%, AcroSeal®
: Thermo Fisher Scientific
Benzonitrile 99+% for spectroscopy
: Thermo Fisher Scientific
Diethyl ether, anhydrous 99+% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
: Thermo Fisher Scientific
Petroleum spirit, 60…90 °C, pure
: CHEMPUR