Order Entry
Poland
ContactUsLinkComponent
HYPERSIL PREP BDS C8 5UM 150X4.6MM COLUMN 1 * 1 items
Catalog # 555-0321
HYPERSIL PREP BDS C8 5UM 150X4.6MM COLUMN 1 * 1 items
Catalog # 555-0321
Frequently Bought Together