Order Entry
Poland
ContactUsLinkComponent
Analtech TLC Uniplates(TM): silica gel matrix, L x W 10 cm x 20 cm , scored, binder, Inorganic, fluorescent indicator 1 * 1 Each
Catalog # SIALZ274283-1PAK
Supplier:  Merck
Analtech TLC Uniplates(TM): silica gel matrix, L x W 10 cm x 20 cm , scored, binder, Inorganic, fluorescent indicator 1 * 1 Each
Catalog # SIALZ274283-1PAK
Supplier:  Merck
Frequently Bought Together