ChromegaChiral(TM) Chiral CCO F4, 3 Micron HPLC Column, 3 mum particle size, L x I.D. 15 cm x 2.1 mm 1 * 1 items
SIALES5579
: Merck
ChromegaChiral(TM) Chiral CCO F4, 3 Micron HPLC Column, 3 mum particle size, L x I.D. 15 cm x 2.1 mm 1 * 1 items
SIALES5579
: Merck
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection